CAS RN 79630-23-2
IUPAC Name:
3-bromo-4-fluorobenzonitrile
Synonyms:
3-bromo-4-fluorobenzonitrile
Molecular Weight (g/mol):
200.01
Molecular Formula:
C7H3BrFN
InChi Key:
JKCYKISVUIVZCS-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1Br)C#N
1
–
1
of
1
results
1
3-Bromo-4-fluorobenzonitrile, 98%
CAS: 79630-23-2 Molecular Formula: C7H3BrFN Molecular Weight (g/mol): 200.01 MDL Number: MFCD00055432 InChI Key: JKCYKISVUIVZCS-UHFFFAOYSA-N Synonym: 5-cyano-2-fluorobromobenzene,benzonitrile, 3-bromo-4-fluoro,3-bromo-4-fluorobenzenecarbonitrile,4-fluoro-3-bromobenzonitrile,3-bromo-4-fluoro-benzonitrile,pubchem1580,3-bromo-4-flurobenzonitrile,acmc-1bh34,3-bromo,4-fluorobenzonitrile,ksc494e4j PubChem CID: 123579 IUPAC Name: 3-bromo-4-fluorobenzonitrile SMILES: C1=CC(=C(C=C1C#N)Br)F