accessibility menu, dialog, popup

UserName

H3COONCH3H3CCH3CH3

CAS RN 79606-43-2

IUPAC Name: 4-methoxy-N,N-bis(propan-2-yl)benzamide
Synonyms: N,N-diisopropyl-4-methoxybenzamide
Molecular Weight (g/mol): 235.33
Molecular Formula: C14H21NO2
InChi Key: IZNMHGFRJFLYSK-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)N(C(C)C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results