accessibility menu, dialog, popup

UserName

CAS RN 79-42-5

IUPAC Name: 2-sulfanylpropanoic acid amine
Synonyms: thiolactic acid
Molecular Weight (g/mol): 123.17
Molecular Formula: C3H9NO2S
InChi Key: VHFSMDHANDABQK-UHFFFAOYNA-N
SMILES: N.CC(S)C(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results