accessibility menu, dialog, popup

UserName

H2NSNH2S

CAS RN 79-40-3

IUPAC Name: ethanedithioamide
Synonyms: dithiooxamide rubeanic acid
Molecular Weight (g/mol): 120.19
Molecular Formula: C2H4N2S2
InChi Key: OAEGRYMCJYIXQT-UHFFFAOYSA-N
SMILES: NC(=S)C(N)=S
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results