accessibility menu, dialog, popup

UserName

CAS RN 79-19-6

H2NNHNH2S

CAS RN 79-19-6

IUPAC Name: aminothiourea
Synonyms: thiosemicarbazide
Molecular Weight (g/mol): 91.13
Molecular Formula: CH5N3S
InChi Key: BRWIZMBXBAOCCF-UHFFFAOYSA-N
SMILES: NNC(N)=S
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)