accessibility menu, dialog, popup

UserName

CAS RN 79-01-6

IUPAC Name: 1,1,2-trichloroethene
Synonyms: trichloroethylene triasol ethinyl trichloride caswell no. 876 trilene TE-141 dow-tri tri-plus M trichlorethylene, tri tri trichlorethylene trichloraethylen, tri
Molecular Weight (g/mol): 131.38
Molecular Formula: C2HCl3
InChi Key: XSTXAVWGXDQKEL-UHFFFAOYSA-N
SMILES: ClC=C(Cl)Cl
Molecular Weight (g/mol) 
  • (19)
  • (1)
Quantity 
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (1)
  • (12)
  • (1)
Physical Form 
  • (14)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (2)
  • (19)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (12)
  • (1)
  • (14)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)