accessibility menu, dialog, popup

UserName

CAS RN 78919-13-8

H3CH3C(S)OH(R)HO(S)(S)OHO(E)(E)

CAS RN 78919-13-8

IUPAC Name: 5-[(2E,3aS,5R,6aS)-5-hydroxy-4-[(1E,3S)-3-hydroxy-4-methyloct-1-en-6-yn-1-yl]-octahydropentalen-2-ylidene]pentanoic acid
Synonyms: iloprost
Molecular Weight (g/mol): 360.49
Molecular Formula: C22H32O4
InChi Key: HIFJCPQKFCZDDL-SGSAMSKHSA-N
SMILES: CC#CCC(C)[C@H](O)\C=C\C1[C@H](O)C[C@@H]2C\C(C[C@H]12)=C/CCCC(O)=O
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (6)
Grade 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (6)