accessibility menu, dialog, popup

UserName

CAS RN 78686-87-0

NClClOCl

CAS RN 78686-87-0

IUPAC Name: 2,5-dichloropyridine-3-carbonyl chloride
Synonyms: 2,5-dichloropyridine-3-carbonyl chloride
Molecular Weight (g/mol): 210.44
Molecular Formula: C6H2Cl3NO
InChi Key: VRJMAKCJTSZUQR-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC(Cl)=CN=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results