accessibility menu, dialog, popup

UserName

CAS RN 78587-05-0

CH3(S)SONONHCl

CAS RN 78587-05-0

IUPAC Name: (4S)-5-(4-chlorophenyl)-N-cyclohexyl-4-methyl-2-oxo-1,3-thiazolidine-3-carboxamide
Synonyms: (4S)-5-(4-chlorophenyl)-N-cyclohexyl-4-methyl-2-oxo-1,3-thiazolidine-3-carboxamide
Molecular Weight (g/mol): 352.88
Molecular Formula: C17H21ClN2O2S
InChi Key: XGWIJUOSCAQSSV-VPHXOMNUSA-N
SMILES: C[C@H]1C(SC(=O)N1C(=O)NC1CCCCC1)C1=CC=C(Cl)C=C1
Solvent or Matrix 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results