accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFFSiClClCl

CAS RN 78560-45-9

IUPAC Name: trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Synonyms: trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Molecular Weight (g/mol): 481.53
Molecular Formula: C8H4Cl3F13Si
InChi Key: PISDRBMXQBSCIP-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results