accessibility menu, dialog, popup

UserName

NH2ClClN

CAS RN 78473-00-4

IUPAC Name: 4-amino-3,5-dichlorobenzonitrile
Synonyms: 4-amino-3,5-dichlorobenzonitrile
Molecular Weight (g/mol): 187.02
Molecular Formula: C7H4Cl2N2
InChi Key: COFNCCWGWXFACE-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C=C1Cl)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results