CAS RN 78-92-2
(+/-)-2-Butanol, 99%
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
sec-Butanol, 99+%, Extra Dry, AcroSeal™
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
sec-Butanol, 99%, Extra Pure
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
sec-Butanol, 99+%, for analysis
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
2-Butanol, anhydrous, 99%
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
2-Butanol, anhydrous, 99%, packaged under Argon in resealable ChemSeal bottles
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
2-Butanol, puriss. p.a., Reag. Ph. Eur., ≥99.5% (GC), Solstice
CAS: 78-92-2 Molecular Formula: C4H10O Molecular Weight (g/mol): 74.12 MDL Number: MFCD00004569 InChI Key: BTANRVKWQNVYAZ-UHFFFAOYNA-N Synonym: 2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol PubChem CID: 6568 ChEBI: CHEBI:35687 IUPAC Name: butan-2-ol SMILES: CCC(C)O
ASTM Method D3606 Check Standard 10-100 mL/L in Isooctane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
ASTM Method D3606 Benzene in Gasoline Kit with IS in Isooctane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
ASTM Method D3606 Check Standard 10-50 mL/L in Isooctane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2-Butanol, Supelco™
CAS: 78-92-2 Grade: Research Molecular Formula: CH3CH(OH)CH2CH3 Molecular Weight (g/mol): 74.12
ASTM Method D3606 Benzene in Gasoline Kit with 10% EtOH & IS in Isooctane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
- 1
- 2