accessibility menu, dialog, popup

UserName

CAS RN 78-79-5

CH3H2CH2C

CAS RN 78-79-5

IUPAC Name: 2-methylbuta-1,3-diene
Synonyms: isoprene isopren 2-methyldivinyl isopreno β-methylbivinyl isopentadiene isoterpene isoprene (substance) isoprene (stabilized)
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: RRHGJUQNOFWUDK-UHFFFAOYSA-N
SMILES: CC(=C)C=C
Molecular Weight (g/mol) 
  • (4)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (1)