accessibility menu, dialog, popup

UserName

CAS RN 78-40-0

CH3OPOOH3COH3C

CAS RN 78-40-0

IUPAC Name: triethyl phosphate
Synonyms: triethyl phosphate TEP triethylphosphate ethyl phosphate
Molecular Weight (g/mol): 182.16
Molecular Formula: C6H15O4P
InChi Key: DQWPFSLDHJDLRL-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)OCC
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (5)
  • (1)
  • (7)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)