accessibility menu, dialog, popup

UserName

CH3OSiOH3COH3COH3C

CAS RN 78-10-4

IUPAC Name: tetraethyl silicate
Synonyms: tetraethyl orthosilicate ethyl polysilicate ethyl silicate
Molecular Weight (g/mol): 208.33
Molecular Formula: C8H20O4Si
InChi Key: BOTDANWDWHJENH-UHFFFAOYSA-N
SMILES: CCO[Si](OCC)(OCC)OCC
Molecular Weight (g/mol) 
  • (12)
  • (9)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (16)
Percent Purity 
  • (2)
  • (2)
  • (10)
  • (1)
Boiling Point 
  • (7)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (12)
  • (9)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (16)
  • (2)
  • (2)
  • (10)
  • (1)
  • (7)
  • (5)