accessibility menu, dialog, popup

UserName

CH3ONONO

CAS RN 7791-49-3

IUPAC Name: 5-methoxy-2,1,3-benzoxadiazol-1-ium-1-olate
Synonyms: 5-methoxy-2,1,3-benzoxadiazol-1-ium-1-olate
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H6N2O3
InChi Key: ABSKHUSUHODMEL-UHFFFAOYSA-N
SMILES: COC1=CC2=NO[N+]([O-])=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results