accessibility menu, dialog, popup

UserName

CaBrBr

CAS RN 7789-41-5

IUPAC Name: calcium dibromide
Synonyms: calcium bromide CaBr2 calcium dibromide kalziumbromid
Molecular Weight (g/mol): 199.89
Molecular Formula: Br2Ca
InChi Key: WGEFECGEFUFIQW-UHFFFAOYSA-L
SMILES: [Ca++].[Br-].[Br-]
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)