CAS RN 7789-23-3
Potassium fluoride, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 SMILES: [F-].[K+]
Potassium fluoride, anhydrous, 99%, Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 SMILES: [F-].[K+]
Potassium fluoride, Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 SMILES: [F-].[K+]
Potassium fluoride, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride kf,potassium fluorure,potassium fluoride, anhydrous,potassiumfluoride,potassium fluoride on calcium fluoride,potassium fluorure french,unii-9082wg1g3f,potassium fluoride,ccris 1568,fluorure de potassium PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium fluoride SMILES: [F-].[K+]
Potassium Fluoride, 99.0+%, Certified AR for Analysis, Anhydrous, Fisher Chemical™
Potassium fluoride, 99.99%, (trace metal basis), Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M IUPAC Name: potassium fluoride SMILES: [F-].[K+]
Potassium fluoride, 40 wt.% on alumina, Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium;fluoride SMILES: [F-].[K+]
Potassium fluoride, ACS, 99% min, Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 SMILES: [F-].[K+]
Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 SMILES: [F-].[K+]
Potassium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium fluoride SMILES: [F-].[K+]
Potassium Fluoride, purum p.a., ≥99.0% (F), Solstice
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium;fluoride SMILES: [F-].[K+]
Potassium fluoride, Purum p.a., ≥99.0% (F), Solstice
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M IUPAC Name: potassium fluoride SMILES: [F-].[K+]
Potassium Fluoride, Supelco™
CAS: 7789-23-3
Potassium Fluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Potassium Fluoride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
- 1
- 2