accessibility menu, dialog, popup

UserName

CsONOO

CAS RN 7789-18-6

IUPAC Name: caesium(1+) nitrate
Synonyms: caesium nitrate
Molecular Weight (g/mol): 194.91
Molecular Formula: CsNO3
InChi Key: NLSCHDZTHVNDCP-UHFFFAOYSA-N
SMILES: [Cs+].[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (15)
  • (3)
Physical Form 
  • (8)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
Grade 
  • (2)
  • (4)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (9)
  • (15)
  • (3)
  • (8)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (1)