accessibility menu, dialog, popup

UserName

CAS RN 7789-00-6

KKOCrOOO

CAS RN 7789-00-6

IUPAC Name: dipotassium dioxochromiumbis(olate)
Synonyms: potassium chromate
Molecular Weight (g/mol): 194.19
Molecular Formula: CrK2O4
InChi Key: XMXNVYPJWBTAHN-UHFFFAOYSA-N
SMILES: [K+].[K+].[O-][Cr]([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
Grade 
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)
  • (2)
  • (8)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (11)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (7)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (8)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)