accessibility menu, dialog, popup

UserName

CAS RN 7787-59-9

HClBiHO

CAS RN 7787-59-9

IUPAC Name: chlorobismuthanone
Synonyms: bismuth oxychloride
Molecular Weight (g/mol): 262.45
Molecular Formula: BiClH2O
InChi Key: OMRKLWMUNWPRFB-UHFFFAOYSA-N
SMILES: Cl.O=[BiH]
Molecular Weight (g/mol) 
  • (3)
  • (8)
Percent Purity 
  • (3)
  • (4)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
Physical Form 
  • (1)
  • (7)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (8)
  • (3)
  • (8)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (7)
  • (3)