CAS RN 7786-30-3
Magnesium chloride, pure
CAS: 7786-30-3 Molecular Formula: Cl2Mg Molecular Weight (g/mol): 95.21 MDL Number: MFCD00011106 InChI Key: TWRXJAOTZQYOKJ-UHFFFAOYSA-L Synonym: magnesium chloride,magnesium chloride anhydrous,mgcl2,magnesium chloride, anhydrous,magnesium 2+ ion dichloride,chloromagnesite,magnesiumchlorid,magnesium ii chloride PubChem CID: 5360315 ChEBI: CHEBI:6636 SMILES: [Mg++].[Cl-].[Cl-]
Magnesium chloride, 1M aq. soln., sterile-filtered
CAS: 7786-30-3 Molecular Formula: Cl2Mg Molecular Weight (g/mol): 95.21 MDL Number: MFCD00011106 InChI Key: TWRXJAOTZQYOKJ-UHFFFAOYSA-L Synonym: magnesium chloride,magnesium chloride anhydrous,mgcl2,magnesium chloride, anhydrous,magnesium 2+ ion dichloride,chloromagnesite,magnesiumchlorid,magnesium ii chloride PubChem CID: 5360315 ChEBI: CHEBI:6636 SMILES: [Mg++].[Cl-].[Cl-]
Magnesium chloride, anhydrous, 99%
CAS: 7786-30-3 Molecular Formula: Cl2Mg Molecular Weight (g/mol): 95.21 MDL Number: MFCD00011106 InChI Key: TWRXJAOTZQYOKJ-UHFFFAOYSA-L Synonym: magnesium chloride,magnesium chloride anhydrous,mgcl2,magnesium chloride, anhydrous,magnesium 2+ ion dichloride,chloromagnesite,magnesiumchlorid,magnesium ii chloride PubChem CID: 5360315 ChEBI: CHEBI:6636 SMILES: [Mg++].[Cl-].[Cl-]
Magnesium chloride, 1M aq. soln.
CAS: 7786-30-3 Molecular Formula: Cl2Mg Molecular Weight (g/mol): 95.21 MDL Number: MFCD00011106 InChI Key: TWRXJAOTZQYOKJ-UHFFFAOYSA-L Synonym: magnesium chloride,magnesium chloride anhydrous,mgcl2,magnesium chloride, anhydrous,magnesium 2+ ion dichloride,chloromagnesite,magnesiumchlorid,magnesium ii chloride PubChem CID: 5360315 ChEBI: CHEBI:6636 SMILES: [Mg++].[Cl-].[Cl-]
Magnesium chloride, ultra dry, 99.9% (metals basis)
CAS: 7786-30-3 Molecular Formula: Cl2Mg Molecular Weight (g/mol): 95.21 MDL Number: MFCD00011106 InChI Key: TWRXJAOTZQYOKJ-UHFFFAOYSA-L Synonym: magnesium chloride,magnesium chloride anhydrous,mgcl2,magnesium chloride, anhydrous,magnesium 2+ ion dichloride,chloromagnesite,magnesiumchlorid,magnesium ii chloride PubChem CID: 5360315 ChEBI: CHEBI:6636 SMILES: [Mg++].[Cl-].[Cl-]
Magnesium chloride, ultra dry, 99.99% (metals basis)
CAS: 7786-30-3 Molecular Formula: Cl2Mg Molecular Weight (g/mol): 95.21 MDL Number: MFCD00011106 InChI Key: TWRXJAOTZQYOKJ-UHFFFAOYSA-L Synonym: magnesium chloride,magnesium chloride anhydrous,mgcl2,magnesium chloride, anhydrous,magnesium 2+ ion dichloride,chloromagnesite,magnesiumchlorid,magnesium ii chloride PubChem CID: 5360315 ChEBI: CHEBI:6636 SMILES: [Mg++].[Cl-].[Cl-]
Magnesium chloride, ultra dry, 99.9% (metals basis)
CAS: 7786-30-3 Molecular Formula: Cl2Mg Molecular Weight (g/mol): 95.21 MDL Number: MFCD00011106 InChI Key: TWRXJAOTZQYOKJ-UHFFFAOYSA-L Synonym: magnesium chloride,magnesium chloride anhydrous,mgcl2,magnesium chloride, anhydrous,magnesium 2+ ion dichloride,chloromagnesite,magnesiumchlorid,magnesium ii chloride PubChem CID: 5360315 ChEBI: CHEBI:6636 IUPAC Name: magnesium;dichloride SMILES: [Mg++].[Cl-].[Cl-]
Magnesium chloride, ultra dry, 99.99% (metals basis)
CAS: 7786-30-3 Molecular Formula: Cl2Mg Molecular Weight (g/mol): 95.21 MDL Number: MFCD00011106 InChI Key: TWRXJAOTZQYOKJ-UHFFFAOYSA-L Synonym: magnesium chloride,magnesium chloride anhydrous,mgcl2,magnesium chloride, anhydrous,magnesium 2+ ion dichloride,chloromagnesite,magnesiumchlorid,magnesium ii chloride PubChem CID: 5360315 ChEBI: CHEBI:6636 SMILES: [Mg++].[Cl-].[Cl-]
Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous
CAS: 7786-30-3 Molecular Formula: Cl2Mg Molecular Weight (g/mol): 95.21 MDL Number: MFCD00011106 InChI Key: TWRXJAOTZQYOKJ-UHFFFAOYSA-L Synonym: magnesium chloride,magnesium chloride anhydrous,mgcl2,magnesium chloride, anhydrous,magnesium 2+ ion dichloride,chloromagnesite,magnesiumchlorid,magnesium ii chloride PubChem CID: 5360315 ChEBI: CHEBI:6636 IUPAC Name: magnesium(2+) dichloride SMILES: [Mg++].[Cl-].[Cl-]
Magnesium Chloride Anhydrous, TRC
CAS: 7786-30-3 Molecular Formula: MgCl2 Molecular Weight (g/mol): 95.21 Synonym: Aerotex Accelerator MX,C-TEK,Catalyst G,Ekimac,FIX-Mg,Freecat MX,HMC 23D,Magnesium Dichloride,Magnesium(II) Chloride,Magnogene,NSC 529832,Soft Wafer,TMT 2,TMT 4,White Nigari,White Nigari NS IUPAC Name: magnesium;dichloride SMILES: [Mg+2].[Cl-].[Cl-]
Melford Magnesium Chloride 2M Solution
Supplied as 5 x 10ml.
Magnesium Chloride, Sigma-Aldrich™
CAS: 7786-30-3
Melford Magnesium Chloride Anhydrous
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- 1
- 2