accessibility menu, dialog, popup

UserName

CAS RN 7784-31-8

H2OH2OH2OH2OH2OH2OH2OH2OH2OAlAlOSOOOOSOOOOSOOO

CAS RN 7784-31-8

IUPAC Name: dialuminium(3+) nonahydrate trisulfate
Synonyms: dialuminium(3+) nonahydrate trisulfate
Molecular Weight (g/mol): 504.27
Molecular Formula: Al2H18O21S3
InChi Key: KFCWRPFNNIEZRY-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (10)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
Grade 
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (5)
  • (3)
Physical Form 
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (1)
  • (1)
  • (5)
  • (3)
  • (1)
  • (5)
  • (10)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)