accessibility menu, dialog, popup

UserName

FFFFZr

CAS RN 7783-64-4

IUPAC Name: zirconium(4+) tetrafluoride
Synonyms: zirconium tetrafluoride
Molecular Weight (g/mol): 167.22
Molecular Formula: F4Zr
InChi Key: OMQSJNWFFJOIMO-UHFFFAOYSA-J
SMILES: [F-].[F-].[F-].[F-].[Zr+4]
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)
  • (4)
  • (1)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)