accessibility menu, dialog, popup

UserName

KKIHHgIHIHHI

CAS RN 7783-33-7

IUPAC Name: dipotassium tetraiodomercurydiuide
Synonyms: nessler's reagent
Molecular Weight (g/mol): 790.44
Molecular Formula: H4HgI4K2
InChi Key: ZCPVFAXYZYZMBC-UHFFFAOYSA-N
SMILES: [K+].[K+].I.I.I.I.[Hg--]
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results