accessibility menu, dialog, popup

UserName

NH4H2OH2OH2OH2OOHPOOONa

CAS RN 7783-13-3

IUPAC Name: ammonium sodium tetrahydrate hydrogen phosphate
Synonyms: microcosmic salt
Molecular Weight (g/mol): 209.07
Molecular Formula: H13NNaO8P
InChi Key: IQTQISLCLWJRPM-UHFFFAOYSA-M
SMILES: [NH4+].O.O.O.O.[Na+].OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (9)
  • (4)
Quantity 
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
Grade 
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (9)
  • (4)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)