accessibility menu, dialog, popup

UserName

CAS RN 7782-86-7

HgONOOHOHOHHONOOHg

CAS RN 7782-86-7

IUPAC Name: mercury(1+) hydrate nitrate
Synonyms: mercury(1+) hydrate nitrate
Molecular Weight (g/mol): 280.61
Molecular Formula: H2HgNO4
InChi Key: UPCSQZXLTBTBCU-UHFFFAOYSA-N
SMILES: O.[Hg+].[O-][N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Grade 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)