accessibility menu, dialog, popup

UserName

C

CAS RN 7782-42-5

IUPAC Name: carbon
Synonyms: carbon kohlenstoff atomic carbon carbonium carbone carbono carbon atom
Molecular Weight (g/mol): 12.01
Molecular Formula: C
InChi Key: OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES: [C]
Quantity 
  • (2)
  • (1)
  • (8)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (6)
  • (7)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (8)
  • (12)
  • (2)
  • (37)
  • (2)
  • (1)
Percent Purity 
  • (5)
  • (2)
  • (24)
  • (1)
Grade 
  • (8)
  • (8)
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (62)
  • (2)
  • (1)
  • (8)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (6)
  • (7)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (8)
  • (12)
  • (2)
  • (37)
  • (2)
  • (1)
  • (5)
  • (2)
  • (24)
  • (1)
  • (8)
  • (8)
  • (4)
  • (4)