accessibility menu, dialog, popup

UserName

CAS RN 77770-09-3

IUPAC Name: 5-iodo-2-methoxyaniline
Synonyms: 5-iodo-2-methoxyaniline
Molecular Weight (g/mol): 249.05
Molecular Formula: C7H8INO
InChi Key: KCAGMESWEMVFON-UHFFFAOYSA-N
SMILES: COC1=C(N)C=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results