accessibility menu, dialog, popup

UserName

CAS RN 7774-34-7

IUPAC Name: calcium hexahydrate dichloride
Synonyms: calcium hexahydrate dichloride
Molecular Weight (g/mol): 219.07
Molecular Formula: CaCl2H12O6
InChi Key: QHFQAJHNDKBRBO-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Ca++]
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (5)
Quantity 
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (4)
Physical Form 
  • (5)
  • (1)
Grade 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (1)
  • (5)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (5)
  • (1)
  • (3)
  • (2)