accessibility menu, dialog, popup

UserName

ClClMn

CAS RN 7773-01-5

IUPAC Name: manganese(2+) tetrahydrate dichloride
Synonyms: manganese(II) chloride
Molecular Weight (g/mol): 197.9
Molecular Formula: Cl2H8MnO4
InChi Key: CNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILES: O.O.O.O.[Cl-].[Cl-].[Mn++]
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (5)
  • (3)
  • (4)
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (6)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)
  • (3)
  • (4)
  • (2)
  • (5)
  • (2)
  • (4)
  • (6)
  • (2)
  • (1)
  • (1)