accessibility menu, dialog, popup

UserName

OOHNNNH2NH2OHO

CAS RN 776277-76-0

IUPAC Name: 6-[(diaminomethylidene)amino]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: 6-[(diaminomethylidene)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 410.47
Molecular Formula: C22H26N4O4
InChi Key: QRELIJBJYZHTQU-UHFFFAOYNA-N
SMILES: NC(N)=NCCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results