accessibility menu, dialog, popup

UserName

KBr

CAS RN 7758-02-3

IUPAC Name: potassium bromide
Synonyms: potassium bromide KBr kaliumbromid
Molecular Weight (g/mol): 119
Molecular Formula: BrK
InChi Key: IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES: [K+].[Br-]
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
Grade 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)