accessibility menu, dialog, popup

UserName

CAS RN 77377-52-7

H3CSiCH3H3CNH3COFFF

CAS RN 77377-52-7

IUPAC Name: N-(butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide
Synonyms: N-(butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide
Molecular Weight (g/mol): 241.33
Molecular Formula: C9H18F3NOSi
InChi Key: NEJIKZHUEMMAEF-UHFFFAOYSA-N
SMILES: CCCC[Si](C)(C)N(C)C(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)