accessibility menu, dialog, popup

UserName

CH3O(R)CH3OH

CAS RN 77301-42-9

IUPAC Name: (1R)-1-(6-methoxynaphthalen-2-yl)ethan-1-ol
Synonyms: (1R)-1-(6-methoxynaphthalen-2-yl)ethanol
Molecular Weight (g/mol): 202.25
Molecular Formula: C13H14O2
InChi Key: OUVJWFRUESFCCY-SECBINFHSA-N
SMILES: COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results