accessibility menu, dialog, popup

UserName

OHClClN

CAS RN 773-76-2

IUPAC Name: 5,7-dichloroquinolin-8-ol
Synonyms: chloroxine chloroxyquinoline quinolor chlorquinol chlorohydroxyquinoline chlofucid quesyl dichlorohydroxyquinoline dichloroxin 哈喹诺
Molecular Weight (g/mol): 214.05
Molecular Formula: C9H5Cl2NO
InChi Key: WDFKMLRRRCGAKS-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (7)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)