accessibility menu, dialog, popup

UserName

BaOSOOO

CAS RN 7727-43-7

IUPAC Name: barium(2+) sulfate
Synonyms: barium sulfate
Molecular Weight (g/mol): 233.38
Molecular Formula: BaO4S
InChi Key: TZCXTZWJZNENPQ-UHFFFAOYSA-L
SMILES: [Ba++].[O-]S([O-])(=O)=O
Quantity 
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
Percent Purity 
  • (4)
  • (4)
  • (6)
  • (3)
Grade 
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
Physical Form 
  • (13)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (9)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (4)
  • (4)
  • (6)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (13)
  • (1)