accessibility menu, dialog, popup

UserName

CH3OOOOH

CAS RN 77269-66-0

IUPAC Name: 3-(2H-1,3-benzodioxol-5-yl)-2-methylpropanoic acid
Synonyms: 3-(2H-1,3-benzodioxol-5-yl)-2-methylpropanoic acid
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: DBXAHUCZEXVUKS-UHFFFAOYNA-N
SMILES: CC(CC1=CC2=C(OCO2)C=C1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results