CAS RN 7718-54-9
Nickel(II) chloride, anhydrous, 98%
CAS: 7718-54-9 Molecular Formula: Cl2Ni Molecular Weight (g/mol): 129.593 MDL Number: MFCD00011142 InChI Key: QMMRZOWCJAIUJA-UHFFFAOYSA-L PubChem CID: 24385 ChEBI: CHEBI:34887 IUPAC Name: dichloronickel SMILES: Cl[Ni]Cl
Nickel(II) chloride, anhydrous, 99.99% (metals basis)
CAS: 7718-54-9 Molecular Formula: Cl2Ni Molecular Weight (g/mol): 129.593 MDL Number: MFCD00011142 InChI Key: QMMRZOWCJAIUJA-UHFFFAOYSA-L PubChem CID: 24385 ChEBI: CHEBI:34887 IUPAC Name: dichloronickel SMILES: Cl[Ni]Cl
Nickel(II) chloride, 98%
CAS: 7718-54-9 MDL Number: MFCD00011142 InChI Key: QMMRZOWCJAIUJA-UHFFFAOYSA-L PubChem CID: 24385 ChEBI: CHEBI:34887
Nickel(II) chloride, anhydrous, 99% (metals basis)
CAS: 7718-54-9 Molecular Formula: Cl2Ni Molecular Weight (g/mol): 129.593 MDL Number: MFCD00011142 InChI Key: QMMRZOWCJAIUJA-UHFFFAOYSA-L PubChem CID: 24385 ChEBI: CHEBI:34887 IUPAC Name: dichloronickel SMILES: Cl[Ni]Cl
Nickel(II) chloride, ultra dry, 99.9% (metals basis)
CAS: 7718-54-9 Molecular Formula: Cl2Ni Molecular Weight (g/mol): 129.593 MDL Number: MFCD00011142 InChI Key: QMMRZOWCJAIUJA-UHFFFAOYSA-L PubChem CID: 24385 ChEBI: CHEBI:34887 IUPAC Name: dichloronickel SMILES: Cl[Ni]Cl
Nickel(II) chloride, 99.99%, (trace metal basis), anhydrous, powder
CAS: 7718-54-9 Molecular Formula: Cl2Ni Molecular Weight (g/mol): 129.6 MDL Number: MFCD00011142 InChI Key: QMMRZOWCJAIUJA-UHFFFAOYSA-L PubChem CID: 24385 ChEBI: CHEBI:34887 IUPAC Name: dichloronickel SMILES: Cl[Ni]Cl
R&D Systems™ DAB Enhancer
R&D Systems™ DAB Enhancer Reagent is a ready-to-use enhancement reagent for DAB chromogens.
Nickel(II) Chloride Anhydrous, TRC
CAS: 7718-54-9 Molecular Formula: NiCl2 Molecular Weight (g/mol): 129.6 IUPAC Name: nickel(2+);dichloride SMILES: [Cl-].[Cl-].[Ni+2]