accessibility menu, dialog, popup

UserName

SOOHNClCl

CAS RN 77167-06-7

IUPAC Name: N-(2,6-dichlorophenyl)benzenesulfonamide
Synonyms: N-(2,6-dichlorophenyl)benzenesulfonamide
Molecular Weight (g/mol): 302.17
Molecular Formula: C12H9Cl2NO2S
InChi Key: CQELSZQJFRLUFQ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1NS(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results