accessibility menu, dialog, popup

UserName

NH2HOOHHCl

CAS RN 770-05-8

IUPAC Name: 4-(2-amino-1-hydroxyethyl)phenol hydrochloride
Synonyms: octopamine hydrochloride
Molecular Weight (g/mol): 189.64
Molecular Formula: C8H12ClNO2
InChi Key: PUMZXCBVHLCWQG-UHFFFAOYNA-N
SMILES: Cl.NCC(O)C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)