CAS RN 77-59-8
CAS RN 77-59-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tomatidine, MedChemExpress
MedChemExpress Tomatidine acts as an anti-inflammatory agent by blocking NF-κB and JNK signaling. Tomatidine activates autophagy either in mammal cells or C elegans.