missing translation for 'onlineSavingsMsg'
Learn More

Tomatidine, MedChemExpress

Product Code. 16489176
Encompass_Preferred
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16489176

Brand: Medchem Express HYN2149

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tomatidine acts as an anti-inflammatory agent by blocking NF-κB and JNK signaling. Tomatidine activates autophagy either in mammal cells or C elegans.

Tomatidine acts as an anti-inflammatory agent by blocking NF-κB and JNK signaling. Tomatidine activates autophagy either in mammal cells or C elegans.

Specifications

Chemical Name or Material Tomatidine
Physical Form Solid
CAS 77-59-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C27H45NO2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 16.67 mg/mL (40.11 mM; ultrasonic and warming and heat to 60°C) ∣0.1 M HCL : < 1 mg/mL (insoluble)
SMILES [H][C@]1(O[C@]2(NC[C@@H](C)CC2)[C@H]3C)C[C@@]4([H])[C@]5([H])CC[C@@]6([H])C[C@@H](O)CC[C@]6(C)[C@@]5([H])CC[C@]4(C)[C@]13[H]
Molecular Weight (g/mol) 415.65
Formula Weight 415.65
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.