accessibility menu, dialog, popup

UserName

CAS RN 7695-91-2

IUPAC Name: (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
Synonyms: tocopheryl acetate tocopherol acetate vitamin E acetate vectan (+-)-α-tocopherol acetate DL-α tocopheryl acetate all rac α-tocopherol acetate α-tocopherol acetate, all rac
Molecular Weight (g/mol): 472.75
Molecular Formula: C31H52O3
InChi Key: ZAKOWWREFLAJOT-CEFNRUSXSA-N
SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (8)
Physical Form 
  • (3)
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (8)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)