CAS RN 7681-76-7
IUPAC Name:
(1-methyl-5-nitro-1H-imidazol-2-yl)methyl carbamate
Synonyms:
ronidazole
Molecular Weight (g/mol):
200.15
Molecular Formula:
C6H8N4O4
InChi Key:
PQFRTXSWDXZRRS-UHFFFAOYSA-N
SMILES:
CN1C(COC(N)=O)=NC=C1[N+]([O-])=O
1
–
4
of
4
results
1
Ronidazole, TRC
CAS: 7681-76-7 Molecular Formula: C6 H8 N4 O4 Molecular Weight (g/mol): 200.15 Synonym: Ronidazole,(1-Methyl-5-nitro-imidazol-2-yl)methyl carbamate IUPAC Name: (1-methyl-5-nitroimidazol-2-yl)methyl carbamate SMILES: Cn1c(COC(=O)N)ncc1[N+](=O)[O-]
2
Ronidazole (1000 ug/mL in Methanol), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Ronidazole, MedChemExpress
MedChemExpress Ronidazole is a potent and orally active antiprotozoal and anti-microbial agent. Ronidazole acts as a veterinary agent against Tritrichomonas foetus in cats models. Ronidazole can be used the research of forhistomoniasis and swine dysentery.