accessibility menu, dialog, popup

UserName

CAS RN 76727-28-1

OOH

CAS RN 76727-28-1

IUPAC Name: (3,4-dihydro-2H-1-benzopyran-3-yl)methanol
Synonyms: 3,4-dihydro-2H-1-benzopyran-3-ylmethanol
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: UPDSWAQIQZFOLF-UHFFFAOYNA-N
SMILES: OCC1COC2=CC=CC=C2C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results