CAS RN 767-58-8
CAS RN 767-58-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methylindan, TRC
CAS: 767-58-8 Molecular Formula: C10H12 Molecular Weight (g/mol): 132.2 Synonym: (RS)-1-Methylindan,1-Methyl-2,3-dihydroindene,1-Methylindan,1-Methylindane,2,3-Dihydro-1-methyl-1H-indene,2,3-Dihydro-1-methyl-1H-indene,NSC 38858 IUPAC Name: 1-methyl-2,3-dihydro-1H-indene SMILES: CC1CCc2ccccc12