accessibility menu, dialog, popup

UserName

NH3

CAS RN 7664-41-7

IUPAC Name: ammonia
Synonyms: ammonia ammonia solution, strong caswell no. 041 E remark 999 am-fol amoniak primary amines ammonia, anhydrous ammoniak kconzentrierter UN 1005 (anhydrous gas or >50% solution)
Molecular Weight (g/mol): 17.03
Molecular Formula: H3N
InChi Key: QGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILES: N
Quantity 
  • (3)
  • (11)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
Solvent or Matrix 
  • (1)
  • (8)
Grade 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (11)
  • (11)
  • (4)
  • (9)
  • (1)
  • (3)
  • (11)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (8)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)