accessibility menu, dialog, popup

UserName

CAS RN 766-97-2

CH3CH

CAS RN 766-97-2

IUPAC Name: 1-ethynyl-4-methylbenzene
Synonyms: 1-ethynyl-4-methylbenzene
Molecular Weight (g/mol): 116.16
Molecular Formula: C9H8
InChi Key: KSZVOXHGCKKOLL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C#C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)