accessibility menu, dialog, popup

UserName

CH3SBr

CAS RN 765-58-2

IUPAC Name: 2-bromo-5-methylthiophene
Synonyms: 2-methyl-5-bromothiophene
Molecular Weight (g/mol): 177.06
Molecular Formula: C5H5BrS
InChi Key: ACDLOOGOFKSUPO-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)S1
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results